Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Antibody detects endogenous levels of total MAK.
The product of this gene is a serine/threonine protein kinase related to kinases involved in cell cycle regulation. It is expressed almost exclusively in the testis, primarily in germ cells. Studies of the mouse and rat homologs have localized the kinase to the chromosomes during meiosis in spermatogenesis, specifically to the synaptonemal complex that exists while homologous chromosomes are paired. There is, however, a study of the mouse homolog that has identified high levels of expression in developing sensory epithelia so its function may be more generalized.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dJ417M14.2; Male germ cell-associated kinase; male germ cell-associated kinase retinal-enriched isoform; Protein kinase RCK; serine/threonine protein kinase MAK; Serine/threonine-protein kinase MAK; testicular secretory protein Li 28
Gene Aliases: A930010O05Rik; MAK; Rck; RP62
UniProt ID: (Human) P20794, (Mouse) E9Q4B2, (Rat) P20793
Entrez Gene ID: (Human) 4117, (Mouse) 17152, (Rat) 25677
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support