Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • MAN2A1 Antibodies

          Invitrogen

          MAN2A1 Polyclonal Antibody

          View all (8) MAN2A1 antibodies

          Promotion

          View available promotion(s)

          Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite MAN2A1 Polyclonal Antibody

          • Antibody Testing Data (1)
          MAN2A1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          MAN2A1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          MAN2A1 Antibody (PA5-120076) in WB

          Western blot analysis of MAN2A1 in extracts of various cell lines. Samples were incubated with MAN2A1 Polyclonal antibody (Product # PA5-120076) using a dilution of 1:1,000, followed by HRP Goat Anti-Rabbit IgG (H+L) at a dilution of 1:10,000. Lysates/proteins: 25 µg per lane. Blocking buffer: 3% nonfat dry milk in TBST. Detection: ECL Basic Kit. Exposure time: 1s. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          MAN2A1 Antibody in Western Blot (WB)

          Product Details

          PA5-120076

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:2,000
          -

          ELISA (ELISA)

          1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant fusion protein containing a sequence corresponding to amino acids 865-1144 of human MAN2A1 (NP_002363.2).
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          2.07 mg/mL

          Purification

          Affinity Chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2913648

          Product Specific Information

          Positive test controls include: LO2, MCF7, NCI-H460, HT-29, HT-1080, Mouse liver, Rat liver. The target is usually found in the following locations: Golgi apparatus membrane, Single-pass type II membrane protein.

          Immunogen sequence: SNIVDIRKVY NREIAMKISS DIKSQNRFYT DLNGYQIQPR MTLSKLPLQA NVYPMTTMAY IQDAKHRLTL LSAQSLGVSS LNSGQIEVIM DRRLMQDDNR GLEQGIQDNK ITANLFRILL EKRSAVNTEE EKKSVSYPSL LSHITSSLMN HPVIPMANKF SSPTLELQGE FSPLQSSLPC DIHLVNLRTI QSKVGNGHSN EAALILHRKG FDCRFSSKGT GLFCSTTQGK ILVQKLLNKF IVESLTPSSL SLMHSPPGTQ NISEINLSPM EISTFRIQLR

          Target Information

          This gene encodes a glycosyl hydrolase that localizes to the Golgi and catalyzes the final hydrolytic step in the asparagine-linked oligosaccharide (N-glycan) maturation pathway. Mutations in the mouse homolog of this gene have been shown to cause a systemic autoimmune disease similar to human systemic lupus erythematosus.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Alpha-mannosidase 2; alpha-mannosidase II; AMan II; Golgi alpha-mannosidase II; golgi integral membrane protein 7; MAN II; mannosidase 2, alpha 1; mannosidase alpha 2; Mannosidase alpha class 2A member 1; mannosidase, alpha type II; mannosidase, alpha, class 2A, member 1; Mannosyl-oligosaccharide 1,3-1,6-alpha-mannosidase

          View more View less

          Gene Aliases: AMan II; GOLIM7; MAN2A1; Mana-2; MANA2; MANII; Manno; Map-2

          View more View less

          UniProt ID: (Human) Q16706, (Mouse) P27046, (Rat) P28494

          View more View less

          Entrez Gene ID: (Human) 4124, (Mouse) 17158, (Rat) 25478

          View more View less

          Function(s)
          mannosyl-oligosaccharide 1,3-1,6-alpha-mannosidase activity zinc ion binding hydrolase activity, hydrolyzing N-glycosyl compounds carbohydrate binding catalytic activity hydrolase activity, hydrolyzing O-glycosyl compounds alpha-mannosidase activity mannosidase activity hydrolase activity hydrolase activity, acting on glycosyl bonds metal ion binding
          Process(es)
          in utero embryonic development liver development carbohydrate metabolic process mannose metabolic process N-glycan processing mitochondrion organization vacuole organization respiratory gaseous exchange metabolic process lung alveolus development positive regulation of neurogenesis retina morphogenesis in camera-type eye protein glycosylation protein deglycosylation
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5d94444789-f7m9l:80/100.66.129.252:80.
          git-commit: ec3573d8beaf0fd412f2d0ea2dcb8dd14c74098c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0