Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: KSLEDVVID IQSSLSKGIR GNEPIYTSTQ EDCINSCCST KNISGDKACN LMIFDTRKTA RQPNCYLFFC PNEEACPLKP AKGLMSYRII TDFPSLTRNL PSQELPQEDS LLHGQFSQAV TPLAHHHTDY SKPTDISWRD TLSQKFGSSD HLEKLFKMDE ASAQLLAYKE KGHSQSSQFS SDQEIAHLLP ENVSALPATV AVASPHTTSA TPKPATLLPT NASVTPSGTS QPQLATTAPP VTTVTSQPPT TLISTVFTRA AATLQAMATT AVLTTTFQAP TDSKGSLETI PFTEISNLTL NTGNVYNPTA LSMSNVESST MNKTASWEGR EASPGSSSQG SVPENQYGLP FE (32-382 aa encoded by BC032998)
MANSC1 gene ontology (GO) annotation includes transmembrane transporter activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Loss of heterozygosity 12 chromosomal region 3 protein; MANSC domain-containing protein 1
Gene Aliases: 9130403P13Rik; LOH12CR3; MANSC1; UNQ316/PRO361
UniProt ID: (Human) Q9H8J5
Entrez Gene ID: (Human) 54682
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support