Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Osenses
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Reconstitute in 150 µL of sterile water. Centrifuge to remove any insoluble material.
Specificity of this antibody: MAP1LC3C.
MAP1LC3C, also named as LC3C, belongs to the MAP1 LC3 family. It is the human homolog of yeast Atg8 and is involved in the formation of autophagosomal vacuoles, called autophagosomes. LC3C is produced by the proteolytic cleavage after the conserved C-terminal Gly residue. The antibody is specific to the prepeptide of LC3C.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Autoph; Autophagy-related protein LC3 C; Autophagy-related ubiquitin-like modifier LC3 C; LC3-like protein 2; MAP1 light chain 3-like protein 2; MAP1 light chain 3-like protein 3; MAP1A/MAP1B LC3 C; MAP1A/MAP1B light chain 3 C; Microtubule-associated protein 1 light chain 3 gamma; Microtubule-associated proteins 1A/1B light chain 3C
Gene Aliases: ATG8J; LC3C; MAP1LC3C
UniProt ID: (Human) Q9BXW4
Entrez Gene ID: (Human) 440738
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support