Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MANFQEHLSC SSSPHLPFSE SKTFNGLQDE LTAMGNHPSP KLLEDQQEKG MVRTELIESV HSPVTTTVLT SVSEDSRDQF ENSVLQLREH DESETAVSQG NSNTVDGEST SGTEDIKIQF SRSGSGSGGF LEGLFGCLRP VWNIIGKAYS TDYKLQQQDT WEVPFEEISE LQWLGSGAQG AVFLGKFRAE EVAIKKVREQ NETDIKHLRK LKHPNIIAFK GVCTQAPCYC IIMEYCAHGQ LYEVLRAGRK ITPRLLVDWS TGIASGMNYL HLHKIIHRDL KSPNVLVTHT DAVKISDFGT SKELSDKSTK MSFAGTVAWM APEVIRNEPV SEKVDIWSFG VVLWELLTGE IPYKDVDSSA IIWGVGSNSL HLPVPSTCPD GFKILMKQTW QSKPRNRPSF RQTLMHLDIA SADVLATPQE TYFKSQAEWR EEVKKHFEKI KSEGTCIHRL DEELIRRRRE ELRHALDIRE HYERKLERAN NLYMELSAIM LQLEMREKEL IKREQAVEKK YPGTYKRHPV RPIIHPNAME KLMKRKGVPH KSGMQTKRPD LLRSEGIPTT EVAPTASPLS GSPKMSTSSS KSRYRSKPRH RRGNSRGSHS DFAAILKNQP AQENSPHPTY LHQAQSQYPS LHHHNSLQQQ YQQPPPAMSQ SHHPRLNMHG QDIATCANNL RYFGPAAALR SPLSNHAQRQ LPGSSPDLIS TAMAADCWRS SEPDKGQAGP WGCCQADAYD PCLQCRPEQY GSLDIPSAEP VGRSPDLSKS PAHNPLLENA QSSEKTEENE FSGCRSESSL GTSHLGTPPA LPRKTRPLQK SGDDSSEEEE GEVDSEVEFP RRQRPHRCIS SCQSYSTFSS ENFSVSDGEE GNTSDHSNSP DELADKLEDR LAEKLDDLLS QTPEIPIDIS SHSDGLSDKE CAVRRVKTQM SLGKLCVEER GYENPMQFEE SDCDSSDGEC SDATVRTNKH YSSATW
Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains. The tyrosine-like kinase (TLK) group consists of 40 tyrosine and serine-threonine kinases such as MLK (mixed-lineage kinase), LISK (LIMK/TESK), IRAK (interleukin-1 receptor-associated kinase), Raf, RIPK (receptor-interacting protein kinase), and STRK (activin and TGF-beta receptors) families.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Leucine zipper-bearing kinase; MAP3K13; Mitogen-activated protein kinase kinase kinase 13; Mixed lineage kinase; MLK
Gene Aliases: LZK; MAP3K13; MEKK13; MLK
UniProt ID: (Human) O43283
Entrez Gene ID: (Human) 9175
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support