Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MAVMEMACPG APGSAVGQQK ELPKAKEKTP PLGKKQSSVY KLEAVEKSPV FCGKWEILND VITKGTAKEG SEAGPAAISI IAQAECENSQ EFSPTFSERI FIAGSKQYSQ SESLDQIPNN VAHATEGKMA RVCWKGKRRS KARKKRKKKS SKSLAHAGVA LAKPLPRTPE QESCTIPVQE DESPLGAPYV RNTPQFTKPL KEPGLGQLCF KQLGEGLRPA LPRSELHKLI SPLQCLNHVW KLHHPQDGGP LPLPTHPFPY SRLPHPFPFH PLQPWKPHPL ESFLGKLACV DSQKPLPDPH LSKLACVDSP KPLPGPHLEP SCLSRGAHEK FSVEEYLVHA LQGSVSSGQA HSLTSLAKTW AARGSRSREP SPKTEDNEGV LLTEKLKPVD YEYREEVHWA THQLRLGRGS FGEVHRMEDK QTGFQCAVKK VRLEVFRAEE LMACAGLTSP RIVPLYGAVR EGPWVNIFME LLEGGSLGQL VKEQGCLPED RALYYLGQAL EGLEYLHSRR ILHGDVKADN VLLSSDGSHA ALCDFGHAVC LQPDGLGKSL LTGDYIPGTE THMAPEVVLG RSCDAKVDVW SSCCMMLHML NGCHPWTQFF RGPLCLKIAS EPPPVREIPP SCAPLTAQAI QEGLRKEPIH RVSAAELGGK VNRALQQVGG LKSPWRGEYK EPRHPPPNQA NYHQTLHAQP RELSPRAPGP RPAEETTGRA PKLQPPLPPE PPEPNKSPPL TLSKEESGMW EPLPLSSLEP APARNPSSPE RKATVPEQEL QQLEIELFLN SLSQPFSLEE QEQILSCLSI DSLSLSDDSE KNPSKASQSS RDTLSSGVHS WSSQAEARSS SWNMVLARGR PTDTPSYFNG VKVQIQSLNG EHLHIREFHR VKVGDIATGI SSQIPAAAFS LVTKDGQPVR YDMEVPDSGI DLQCTLAPDG SFAWSWRVKH GQLENRP
MAP3K14 (NIK) is structurally related to MAP3Ks. MAP3K14 phosphorylates and activates CHUK (IKKalpha) and IKBKB (IKK beta), which in turn phosphorylate IkB-alpha, releasing NF-KappaB leading to up-regulation of inflammatory response genes. This kinase binds to TRAF2 and stimulates NF-kappaB activity. It shares sequence similarity with several other MAPKK kinases. It participates in an NF-kappaB-inducing signalling cascade common to receptors of the tumour-necrosis/nerve-growth factor (TNF/NGF) family and to the interleukin-1 type-I receptor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HsNIK; Mitogen-activated protein kinase kinase kinase 14; NF-kappa-beta-inducing kinase; Serine/threonine-protein kinase NIK
Gene Aliases: FTDCR1B; HS; HSNIK; MAP3K14; NIK
UniProt ID: (Human) Q99558
Entrez Gene ID: (Human) 9020
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support