Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • ASK1 Antibodies

          Abnova

          MAP3K5 Polyclonal Antibody, MaxPab™

          View all (51) ASK1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite MAP3K5 Polyclonal Antibody, MaxPab™

          • Antibody Testing Data (2)
          MAP3K5 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          MAP3K5 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          MAP3K5 Antibody (H00004217-D01P) in WB

          Western Blot analysis of MAP3K5 expression in transfected 293T cell line (H00004217-T01) by MAP3K5 MaxPab polyclonal antibody.

          Lane 1: MAP3K5 transfected lysate(154.50 KDa).
          Lane 2: Non-transfected lysate.
          {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          MAP3K5 Antibody in Western Blot (WB)
          MAP3K5 Antibody in Proximity Ligation Assay (PLA) (PLA)

          Product Details

          H00004217-D01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-5 µg/mL
          -

          in situ PLA (PLA)

          1:1200
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          MAP3K5 (NP_005914.1, 1 a.a. approximately 1374 a.a) full-length human protein.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          See Label

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Sequence of this protein is as follows: MSTEADEGIT FSVPPFAPSG FCTIPEGGIC RRGGAAAVGE GEEHQLPPPP PGSFWNVESA AAPGIGCPAA TSSSSATRGR GSSVGGGSRR TTVAYVINEA SQGQLVVAES EALQSLREAC ETVGATLETL HFGKLDFGET TVLDRFYNAD IAVVEMSDAF RQPSLFYHLG VRESFSMANN IILYCDTNSD SLQSLKEIIC QKNTMCTGNY TFVPYMITPH NKVYCCDSSF MKGLTELMQP NFELLLGPIC LPLVDRFIQL LKVAQASSSQ YFRESILNDI RKARNLYTGK ELAAELARIR QRVDNIEVLT ADIVINLLLS YRDIQDYDSI VKLVETLEKL PTFDLASHHH VKFHYAFALN RRNLPGDRAK ALDIMIPMVQ SEGQVASDMY CLVGRIYKDM FLDSNFTDTE SRDHGASWFK KAFESEPTLQ SGINYAVLLL AAGHQFESSF ELRKVGVKLS SLLGKKGNLE KLQSYWEVGF FLGASVLAND HMRVIQASEK LFKLKTPAWY LKSIVETILI YKHFVKLTTE QPVAKQELVD FWMDFLVEAT KTDVTVVRFP VLILEPTKIY QPSYLSINNE VEEKTISIWH VLPDDKKGIH EWNFSASSVR GVSISKFEER CCFLYVLHNS DDFQIYFCTE LHCKKFFEMV NTITEEKGRS TEEGDCESDL LEYDYEYDEN GDRVVLGKGT YGIVYAGRDL SNQVRIAIKE IPERDSRYSQ PLHEEIALHK HLKHKNIVQY LGSFSENGFI KIFMEQVPGG SLSALLRSKW GPLKDNEQTI GFYTKQILEG LKYLHDNQIV HRDIKGDNVL INTYSGVLKI SDFGTSKRLA GINPCTETFT GTLQYMAPEI IDKGPRGYGK AADIWSLGCT IIEMATGKPP FYELGEPQAA MFKVGMFKVH PEIPESMSAE AKAFILKCFE PDPDKRACAN DLLVDEFLKV SSKKKKTQPK LSALSAGSNE YLRSISLPVP VLVEDTSSSS EYGSVSPDTE LKVDPFSFKT RAKSCGERDV KGIRTLFLGI PDENFEDHSA PPSPEEKDSG FFMLRKDSER RATLHRILTE DQDKIVRNLM ESLAQGAEEP KLKWEHITTL IASLREFVRS TDRKIIATTL SKLKLELDFD SHGISQVQVV LFGFQDAVNK VLRNHNIKPH WMFALDSIIR KAVQTAITIL VPELRPHFSL ASESDTADQE DLDVEDDHEE QPSNQTVRRP QAVIEDAVAT SGVSTLSSTV SHDSQSAHRS LNVQLGRMKI ETNRLLEELV RKEKELQALL HRAIEEKDQE IKHLKLKSQP IEIPELPVFH LNSSGTNTED SELTDWLRVN GADEDTISRF LAEDYTLLDV LYYVTRDDLK CLRLRGGMLC TLWKAIIDFR NKQT

          Target Information

          MAP3K5 (ASK1) is a serine/threonine kinase that is abundantly expressed in human heart and pancreas and are activated in response to various extracellular stimuli, including cytokines, growth factors and environmental stresses. A novel MAP kinase kinase kinase (MAPKKK) was identified and designated ASK1 (for apoptosis signal-regulating kinase 1) and MAPKKK5. ASK1 activates two different subgroups of MAPKK, MKK4 and MKK6, which in turn activates c-Jun N-terminal kinase (JNK) and p38 MAP kinase, respectively. ASK1/MAPKKK5 is activated by TNFR and Fas through the interaction with members of the TRAF family and Fas-associated protein Daxx. Overexpression of ASK1 induced apoptotic cell death and a catalytically inactive form of ASK1 inhibited TNF-a-induced apoptosis. ASK1 is expressed in variety of human and mouse tissues.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: apoptosis signal regulating kinase 1; Apoptosis signal-regulating kinase 1; ASK 1; ASK-1; EC 2.7.11.25; kinase ASK1; M3K5; MAP/ERK kinase kinase 5; MAPK/ERK kinase kinase 5; MEK kinase 5; MEKK 5; Mitogen-activated protein kinase kinase kinase 5; RP3-325F22.4

          View more View less

          Gene Aliases: ASK1; MAP3K5; MAPKKK5; MEKK5

          View more View less

          UniProt ID: (Human) Q99683

          View more View less

          Entrez Gene ID: (Human) 4217

          View more View less

          Function(s)
          magnesium ion binding protein kinase activity protein serine/threonine kinase activity MAP kinase kinase kinase activity protein binding ATP binding protein kinase binding protein phosphatase binding protein homodimerization activity
          Process(es)
          MAPK cascade activation of MAPKK activity response to ischemia protein phosphorylation JNK cascade activation of JUN kinase activity intrinsic apoptotic signaling pathway in response to oxidative stress viral process response to endoplasmic reticulum stress p38MAPK cascade positive regulation of apoptotic process positive regulation of cysteine-type endopeptidase activity involved in apoptotic process positive regulation of JUN kinase activity innate immune response positive regulation of myoblast differentiation positive regulation of JNK cascade intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress cellular response to hydrogen peroxide apoptotic signaling pathway programmed necrotic cell death positive regulation of neuron death cellular response to reactive nitrogen species
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0