Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • MARCH10 Antibodies

          Abnova

          MARCH10 Polyclonal Antibody, MaxPab™

          View all (7) MARCH10 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite MARCH10 Polyclonal Antibody, MaxPab™

          • Antibody Testing Data (2)
          MARCH10 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          MARCH10 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          MARCH10 Antibody (H00162333-D01P) in WB

          Western Blot analysis of MARCH10 expression in transfected 293T cell line (H00162333-T02) by MARCH10 MaxPab polyclonal antibody.

          Lane 1: MARCH10 transfected lysate(73.50 KDa).
          Lane 2: Non-transfected lysate.
          {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          MARCH10 Antibody in Western Blot (WB)
          MARCH10 Antibody in Western Blot (WB)

          Product Details

          H00162333-D01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          MARCH10 (AAH35021.1, 1 a.a. approximately 657 a.a) full-length human protein.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          See Label

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Sequence of this protein is as follows: MLHDARDRQK FFSDVQYLRD MQHKVDSEYQ ACLRRQEYRR DPNEKKRDQF WGQETSFERS RFSSRSSSKQ SSSEEDALTE PRSSIKISAF KCDSKLPAID QTSVKQKHKS TMTVRKAEKV DPSEPSPADQ APMVLLRKRK PNLRRFTVSP ESHSPRASGD RSRQKQQWPA KVPVPRGADQ VVQQEGLMCN TKLKRPNQER RNLVPSSQPM TENAPDRAKK GDPSAPSQSE LHPALSQAFQ EKNSPQVLSE FSGPPLTPTT VGGPRKASFR FRDEDFYSIL SLNSRRESDD TEEETQSEEC LWVGVRSPCS PSHHKRSRFG GTSTPQAKNK NFEENAENCR GHSSRRSEPS HGSLRISNAM EPATERPSAG QRLSQDPGLP DRESATEKDR GGSENAKKSP LSWDTKSEPR QEVGVNAENV WSDCISVEHR PGTHDSEGYW KDYLNSSQNS LDYFISGRPI SPRSSVNSSY NPPASFMHSA LRDDIPVDLS MSSTSVHSSD SEGNSGFHVC QPLSPIRNRT PFASAENHNY FPVNSAHEFA VREAEDTTLT SQPQGAPLYT DLLLNPQGNL SLVDSSSSSP SRMNSEGHLH VSGSLQENTP FTFFAVSHFP NQNDNGSRMA ASGFTDEKET SKIKADPEKL KKLQESLLEE DSEEEGD

          Target Information

          MARCH10 is a member of the MARCH family of membrane-bound E3 ubiquitin ligases to target lysines in substrate proteins, thereby signaling their vesicular transport between membrane compartments.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 10-Mar; FLJ35757; MARCH-X; membrane associated ring finger 10; membrane-associated ring finger (C3HC4) 10, E3 ubiquitin protein ligase; Membrane-associated RING finger protein 10; Membrane-associated RING-CH protein X; probable E3 ubiquitin-protein ligase MARCH10; Probable E3 ubiquitin-protein ligase MARCHF10; RING finger protein 190; RING-type E3 ubiquitin transferase MARCHF10; testis secretory sperm-binding protein Li 228n

          View more View less

          Gene Aliases: 4933417C16Rik; AV039223; MARCH-X; MARCH10; MARCHF10; OTTMUSG00000002847; RNF190

          View more View less

          UniProt ID: (Human) Q8NA82

          View more View less

          Entrez Gene ID: (Human) 162333, (Mouse) 632687

          View more View less

          Function(s)
          protein binding zinc ion binding ligase activity molecular_function
          Process(es)
          protein ubiquitination biological_process
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-bff4f7d6d-tmrcx:80/100.66.135.11:80.
          git-commit: 683fd26f943b2dde6a08880618459b58d20c3896
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-Offline