Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
MARCH2 is a member of the MARCH family of membrane-bound E3 ubiquitin ligases (EC 6.3.2.19). MARCH enzymes add ubiquitin (see MIM 191339) to target lysines in substrate proteins, thereby signaling their vesicular transport between membrane compartments. MARCH2 reduces surface accumulation of several glycoproteins and appears to regulate early endosome-to-trans-Golgi network (TGN) trafficking (Bartee et al., 2004 [PubMed 14722266]; Nakamura et al., 2005 [PubMed 15689499]).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 ubiquitin-protein ligase MARCH2; E3 ubiquitin-protein ligase MARCHF2; MARCH-II; membrane associated ring finger 2; membrane associated RING-CH finger protein II; membrane-associated ring finger (C3HC4) 2, E3 ubiquitin protein ligase; Membrane-associated RING finger protein 2; Membrane-associated RING-CH protein II; RING finger protein 172; RING-type E3 ubiquitin transferase MARCHF2; RNF172; syntaxin-6-binding protein
Gene Aliases: 9530046H09Rik; HSPC240; MARCH-II; MARCH2; MARCHF2; RGD1306395; RNF172
UniProt ID: (Human) Q9P0N8, (Rat) Q5I0I2, (Mouse) Q99M02
Entrez Gene ID: (Human) 51257, (Rat) 362849, (Mouse) 224703
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support