Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: HEGSSVYRIF KRWQAVNQQW KVLNYDKTKD IGGDAGGGTA GKSGPRNSRT GPTSGATSRP PAAQRMRTLL PQRCGYTILH LLGQLRPPDA RSSSHSGREV VMRVTT
MARCH9 is a member of the MARCH family of membrane-bound E3 ubiquitin ligases (EC 6.3.2.19). MARCH enzymes add ubiquitin to target lysines in substrate proteins, thereby signaling their vesicular transport between membrane compartments. MARCH9 induces internalization of several membrane glycoproteins and directs them to the endosomal compartment.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 9-Mar; E3 ubiquitin-protein ligase MARCHF9; MARCH-IX; membrane associated ring finger 9; membrane-associated ring finger (C3HC4) 9; Membrane-associated RING finger protein 9; Membrane-associated RING-CH protein IX; RING finger protein 179; RING-type E3 ubiquitin transferase MARCHF9
Gene Aliases: MARCH-IX; MARCH9; MARCHF9; RNF179
UniProt ID: (Human) Q86YJ5
Entrez Gene ID: (Human) 92979
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support