Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen sequence: MAAATEHNRP SSGDRNLERR CSPNLSREVL YEIFRSLHTL VGQLDLRDDV VKITIDWNKL QSLSAFQPAL LFSALEQHIL YLQPFLAKLQ SPIKEENTTA VEEIGRTEMG NKNEVNDKFS IGDLQEEEKH KESDLRDVKK TQIHFDPEVV QIKAGKAEID RRISAFIERK QAEINENNVR EFCNVIDCNQ ENSCARTDAI FTPYPGFKSH VKVSRVVYTY GPQTRPEGIP GSGHKPNSML RDCGNQAVEE RLQNIEAHLR LQTGGPVPRD IYQRIKKLED KILELEGISP EYFQSVSFSG KRRKVQPPQQ NYSLAELDEK ISALKQALLR KSREAESMAT HHLP
MBIP inhibits the MAP3K12 activity to induce the activation of the JNK/SAPK pathway. Component of the ATAC complex, a complex with histone acetyltransferase activity on histones H3 and H4.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MAP3K12-binding inhibitory protein 1; MAPK upstream kinase-binding inhibitory protein; MUK-binding inhibitory protein
Gene Aliases: 4933408E06Rik; BM-015; MBIP
UniProt ID: (Human) Q9NS73, (Mouse) Q99LQ1
Entrez Gene ID: (Human) 51562, (Mouse) 217588
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support