Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: ENGRVIACFD SLKGRCSREN CKYLHPPTHL KTQLEINGRN NLIQQKTAAA
Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%
This gene encodes a C3H-type zinc finger protein, which is similar to the Drosophila melanogaster muscle blind B protein. Drosophila muscle blind is a gene required for photoreceptor differentiation. Several alternatively spliced transcript variants have been described but the full-length natures of only some have been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp781H1296; MGC120625; MGC120626; MGC120628; muscleblind-like 2; Muscleblind-like protein 1; Muscleblind-like protein 2; Muscleblind-like protein-like; Muscleblind-like protein-like 39; muscleblind-like splicing regulator 2; RP11-128N14.1
Gene Aliases: MBLL; MBLL39; MBNL2; MLP1; PRO2032
UniProt ID: (Human) Q5VZF2
Entrez Gene ID: (Human) 10150
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support