Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: SAASAMALQ PGTLQLIPKR SALEKPNGAT PVFNPTVFHC QQALTNLQLP QPAFIPAGPI LCMAPASNIV PMMHGA (156-230 aa encoded by BC042090)
This gene encodes a member of the muscleblind-like family of proteins. The encoded protein may function in regulation of alternative splicing and may play a role in the pathophysiology of myotonic dystrophy. Alternatively spliced transcript variants have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cys3His CCG1-required protein; muscleblind-like 3; Muscleblind-like protein 3; muscleblind-like splicing regulator 3; Muscleblind-like X-linked protein; Protein HCHCR; Protein MCHCR; RP5-842K24.3
Gene Aliases: A530038J18Rik; AI661274; CHCR; E430034C16Rik; MBLX; MBLX39; MBNL3; MBXL
UniProt ID: (Human) Q9NUK0, (Mouse) Q8R003
Entrez Gene ID: (Human) 55796, (Mouse) 171170
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support