Product References
The Valsa Mali effector Vm1G-1794 protects the aggregated MdEF-Tu from autophagic degradation to promote infection in apple.
Autophagy
Che R,Liu C,Wang Q,Tu W,Wang P,Li C,Gong X,Mao K,Feng H,Huang L,Li P,Ma F
Thu Jun 01 00:00:00 UTC 2023
In vitro methylation of the U7 snRNP subunits Lsm11 and SmE by the PRMT5/MEP50/pICln methylosome.
bioRxiv : the preprint server for biology
Yang XC,Desotell A,Lin MH,Paige AS,Malinowska A,Sun Y,Aik WS,Dadlez M,Tong L,Dominski Z
Wed May 10 00:00:00 UTC 2023
De novo variants in MRTFB have gain-of-function activity in Drosophila and are associated with a novel neurodevelopmental phenotype with dysmorphic features.
Genetics in medicine : official journal of the American College of Medical Genetics
Andrews JC,Mok JW,Kanca O,Jangam S,Tifft C,Macnamara EF,Russell BE,Wang LK,Nelson SF,Bellen HJ,Yamamoto S,Malicdan MCV,Wangler MF
Thu Jun 01 00:00:00 UTC 2023
Structural mechanism of BRD4-NUT and p300 bipartite interaction in propagating aberrant gene transcription in chromatin in NUT carcinoma.
Nature communications
Yu D,Liang Y,Kim C,Jaganathan A,Ji D,Han X,Yang X,Jia Y,Gu R,Wang C,Zhang Q,Cheung KL,Zhou MM,Zeng L
Tue Jan 24 00:00:00 UTC 2023
Screening and identification of cyprinid herpesvirus 2 (CyHV-2) ORF55-interacting proteins by phage display.
Virology journal
Qian M,Xiao S,Yang Y,Yu F,Wen J,Lu L,Wang H
Wed Apr 12 00:00:00 UTC 2023
Structural insights into pathogenic mechanism of hypohidrotic ectodermal dysplasia caused by ectodysplasin A variants.
Nature communications
Yu K,Huang C,Wan F,Jiang C,Chen J,Li X,Wang F,Wu J,Lei M,Wu Y
Sat Feb 11 00:00:00 UTC 2023
Activation and negative feedback regulation of SlHY5 transcription by the SlBBX20/21-SlHY5 transcription factor module in UV-B signaling.
The Plant cell
Yang G,Zhang C,Dong H,Liu X,Guo H,Tong B,Fang F,Zhao Y,Yu Y,Liu Y,Lin L,Yin R
Tue Apr 26 00:00:00 UTC 2022
Mining and characterization of oxidative stress-related binding proteins of parthenolide in Xanthomonas oryzae pv. oryzae.
Pest management science
Zhou Q,Li L,Liu F,Hu J,Cao Y,Qiao S,Zhou Y,Wang B,Jia Y,Chen Y,Xu S,Feng X
Mon Aug 01 00:00:00 UTC 2022
Insights into acylation mechanisms: co-expression of serine carboxypeptidase-like acyltransferases and their non-catalytic companion paralogs.
The Plant journal : for cell and molecular biology
Yao S,Liu Y,Zhuang J,Zhao Y,Dai X,Jiang C,Wang Z,Jiang X,Zhang S,Qian Y,Tai Y,Wang Y,Wang H,Xie DY,Gao L,Xia T
Fri Jul 01 00:00:00 UTC 2022
Amelioration of hepatic steatosis by dietary essential amino acid-induced ubiquitination.
Molecular cell
Zhang Y,Lin S,Peng J,Liang X,Yang Q,Bai X,Li Y,Li J,Dong W,Wang Y,Huang Y,Pei Y,Guo J,Zhao W,Zhang Z,Liu M,Zhu AJ
Thu Apr 21 00:00:00 UTC 2022
Disease-linked TDP-43 hyperphosphorylation suppresses TDP-43 condensation and aggregation.
The EMBO journal
Gruijs da Silva LA,Simonetti F,Hutten S,Riemenschneider H,Sternburg EL,Pietrek LM,Gebel J,Dötsch V,Edbauer D,Hummer G,Stelzl LS,Dormann D
Tue Apr 19 00:00:00 UTC 2022
Energy sensor AMPK gamma regulates translation via phosphatase PPP6C independent of AMPK alpha.
Molecular cell
Zhou Q,Hao B,Cao X,Gao L,Yu Z,Zhao Y,Zhu M,Zhong G,Chi F,Dai X,Mao J,Zhu Y,Rong P,Chen L,Bai X,Ye C,Chen S,Liang T,Li L,Feng XH,Tan M,Zhao B
Thu Dec 15 00:00:00 UTC 2022
Non-invasive photobiomodulation treatment in an Alzheimer Disease-like transgenic rat model.
Theranostics
Yang L,Wu C,Parker E,Li Y,Dong Y,Tucker L,Brann DW,Lin HW,Zhang Q
Thu Apr 21 00:00:00 UTC 2022
Regulatory basis for reproductive flexibility in a meningitis-causing fungal pathogen.
Nature communications
Hu P,Ding H,Liu H,Yang Y,Chen L,He GJ,Ke W,Zhu P,Tian X,Peng Y,Shen Z,Yao X,Tao C,Yang E,Liao G,Liu X,Wang L
Sat Dec 24 00:00:00 UTC 2022
The 'Candidatus Phytoplasma tritici' effector SWP12 degrades the transcription factor TaWRKY74 to suppress wheat resistance.
The Plant journal : for cell and molecular biology
Bai B,Zhang G,Li Y,Wang Y,Sujata S,Zhang X,Wang L,Zhao L,Wu Y
Thu Dec 01 00:00:00 UTC 2022
The histone deacetylase 1/GSK3/SHAGGY-like kinase 2/BRASSINAZOLE-RESISTANT 1 module controls lateral root formation in rice.
Plant physiology
Hou J,Zheng X,Ren R,Shi Q,Xiao H,Chen Z,Yue M,Wu Y,Hou H,Li L
Wed Jun 01 00:00:00 UTC 2022
MAPKs trigger antiviral immunity by directly phosphorylating a rhabdovirus nucleoprotein in plants and insect vectors.
The Plant cell
Ding ZH,Gao Q,Tong X,Xu WY,Ma L,Zhang ZJ,Wang Y,Wang XB
Sat Jul 30 00:00:00 UTC 2022
Long-term exercise pre-training attenuates Alzheimer's disease-related pathology in a transgenic rat model of Alzheimer's disease.
GeroScience
Yang L,Wu C,Li Y,Dong Y,Wu CY,Lee RH,Brann DW,Lin HW,Zhang Q
Wed Jun 01 00:00:00 UTC 2022
The RNA-binding protein FUS is chaperoned and imported into the nucleus by a network of import receptors.
The Journal of biological chemistry
Baade I,Hutten S,Sternburg EL,Pörschke M,Hofweber M,Dormann D,Kehlenbach RH
Mon Aug 23 00:00:00 UTC 2021
A Novel Antigenic Site Spanning Domains I and III of the Zika Virus Envelope Glycoprotein Is the Target of Strongly Neutralizing Human Monoclonal Antibodies.
Journal of virology
Graham SD,Tu HA,McElvany BD,Graham NR,Grinyo A,Davidson E,Doranz BJ,Diehl SA,de Silva AM,Markmann AJ
Mon Apr 12 00:00:00 UTC 2021
The missing linker between SUN5 and PMFBP1 in sperm head-tail coupling apparatus.
Nature communications
Zhang Y,Liu C,Wu B,Li L,Li W,Yuan L
Fri Aug 13 00:00:00 UTC 2021
The missing linker between SUN5 and PMFBP1 in sperm head-tail coupling apparatus.
Nature communications
Zhang Y,Liu C,Wu B,Li L,Li W,Yuan L
Fri Aug 13 00:00:00 UTC 2021
Heat shock protein 70, glutamate dehydrogenase, and angiotensin-converting enzyme of Bombyx mori mediate the cell attachment of Cypovirus 1.
The Journal of general virology
Yang J,Wang J,Peng X,Lei C,Sun X,Hu J
Wed Dec 01 00:00:00 UTC 2021
Nod factor receptor complex phosphorylates GmGEF2 to stimulate ROP signaling during nodulation.
Current biology : CB
Gao JP,Xu P,Wang M,Zhang X,Yang J,Zhou Y,Murray JD,Song CP,Wang E
Mon Aug 23 00:00:00 UTC 2021
Functional regulation of an ancestral RAG transposon ProtoRAG by a trans-acting factor YY1 in lancelet.
Nature communications
Liu S,Yuan S,Gao X,Tao X,Yu W,Li X,Chen S,Xu A
Wed Sep 09 00:00:00 UTC 2020
A Quantitative Assay to Measure Stress Granule Association of Proteins and Peptidesin Semi-permeabilized Human Cells.
Bio-protocol
Hutten S,Dormann D
Sun Dec 20 00:00:00 UTC 2020
The Extended C-Terminal Region of Influenza C Virus Nucleoprotein Is Important for Nuclear Import and Ribonucleoprotein Activity.
Journal of virology
Tang YS,Lo CY,Mok CK,Chan PK,Shaw PC
Wed May 01 00:00:00 UTC 2019
Characterization of Ferredoxin-Dependent Biliverdin Reductase PCYA1 Reveals the Dual Function in Retrograde Bilin Biosynthesis and Interaction With Light-Dependent Protochlorophyllide Oxidoreductase LPOR in Chlamydomonas reinhardtii.
Frontiers in plant science
Zhang W,Zhong H,Lu H,Zhang Y,Deng X,Huang K,Duanmu D
Wed Sep 30 00:00:00 UTC 2020
Characterization of Ferredoxin-Dependent Biliverdin Reductase PCYA1 Reveals the Dual Function in Retrograde Bilin Biosynthesis and Interaction With Light-Dependent Protochlorophyllide Oxidoreductase LPOR in Chlamydomonas reinhardtii.
Frontiers in plant science
Zhang W,Zhong H,Lu H,Zhang Y,Deng X,Huang K,Duanmu D
Wed Sep 30 00:00:00 UTC 2020
Unspliced XBP1 Confers VSMC Homeostasis and Prevents Aortic Aneurysm Formation via FoxO4 Interaction.
Circulation research
Zhao G,Fu Y,Cai Z,Yu F,Gong Z,Dai R,Hu Y,Zeng L,Xu Q,Kong W
Fri Dec 08 00:00:00 UTC 2017
Unspliced XBP1 Confers VSMC Homeostasis and Prevents Aortic Aneurysm Formation via FoxO4 Interaction.
Circulation research
Zhao G,Fu Y,Cai Z,Yu F,Gong Z,Dai R,Hu Y,Zeng L,Xu Q,Kong W
Fri Dec 08 00:00:00 UTC 2017
Matrix Metalloproteinase 9 Facilitates Hepatitis B Virus Replication through Binding with Type I Interferon (IFN) Receptor 1 To Repress IFN/JAK/STAT Signaling.
Journal of virology
Chen J,Xu W,Chen Y,Xie X,Zhang Y,Ma C,Yang Q,Han Y,Zhu C,Xiong Y,Wu K,Liu F,Liu Y,Wu J
Sat Apr 15 00:00:00 UTC 2017
Matrix Metalloproteinase 9 Facilitates Hepatitis B Virus Replication through Binding with Type I Interferon (IFN) Receptor 1 To Repress IFN/JAK/STAT Signaling.
Journal of virology
Chen J,Xu W,Chen Y,Xie X,Zhang Y,Ma C,Yang Q,Han Y,Zhu C,Xiong Y,Wu K,Liu F,Liu Y,Wu J
Sat Apr 15 00:00:00 UTC 2017
SCFAtPP2-B11 modulates ABA signaling by facilitating SnRK2.3 degradation in Arabidopsis thaliana.
PLoS genetics
Cheng C,Wang Z,Ren Z,Zhi L,Yao B,Su C,Liu L,Li X
Tue Aug 01 00:00:00 UTC 2017
SCFAtPP2-B11 modulates ABA signaling by facilitating SnRK2.3 degradation in Arabidopsis thaliana.
PLoS genetics
Cheng C,Wang Z,Ren Z,Zhi L,Yao B,Su C,Liu L,Li X
Tue Aug 01 00:00:00 UTC 2017
A New Member of the TBC1D15 Family from Chiloscyllium plagiosum: Rab GTPase-Activating Protein Based on Rab7 as a Substrate.
Marine drugs
Li Y,Wang W,Cheng D,Wang T,Lu C,Chen J,Nie Z,Zhang W,Lv Z,Wu W,Shu J
Wed May 13 00:00:00 UTC 2015
A New Member of the TBC1D15 Family from Chiloscyllium plagiosum: Rab GTPase-Activating Protein Based on Rab7 as a Substrate.
Marine drugs
Li Y,Wang W,Cheng D,Wang T,Lu C,Chen J,Nie Z,Zhang W,Lv Z,Wu W,Shu J
Wed May 13 00:00:00 UTC 2015