Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SVESPNLTTP ITSNPTDTRK ITSGNSSNSP NAEVMAVQKK LDSIIDLTKE GLSNCNTESP VSPLESHSKA ASNSKETTPL AQNAVQVPES FEHLPPLPEP PAPL
Highest antigen sequence identity to the following orthologs: Mouse - 58%, Rat - 40%.
Recruiter that couples transcriptional factors to general transcription apparatus and thereby modulates transcription regulation and chromatin formation. Can both act as an activator or a repressor depending on the context. Mediates MBD1-dependent transcriptional repression, probably by recruiting complexes containing SETDB1. The complex formed with MBD1 and SETDB1 represses transcription and probably couples DNA methylation and histone H3 'Lys-9' trimethylation (H3K9me3) activity (Probable). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Activating transcription factor 7-interacting protein 2; ATF7-interacting protein 2; MBD1-containing chromatin associated factor 2; MBD1-containing chromatin-associated factor 2
Gene Aliases: ATF7IP2; MCAF2
UniProt ID: (Human) Q5U623
Entrez Gene ID: (Human) 80063
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support