Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
This gene is similar to the mitochondrial carrier triple repeat 1 gene on chromosome 9. The gene is intronless and may be an evolving pseudogene; however, it is transcribed and it contains a full-length coding region so it is currently classified as a protein-coding locus.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: mitochondrial carrier triple repeat 2; Mitochondrial carrier triple repeat protein 2; Mitochondrial NAD(+) transporter SLC25A52; Mitochondrial nicotinamide adenine dinucleotide transporter SLC25A52; Solute carrier family 25 member 52; solute carrier family 25, member 52
Gene Aliases: MCART2; SLC25A52
UniProt ID: (Human) Q3SY17
Entrez Gene ID: (Human) 147407
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support