Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: GTELAETELP NDVQSTSSVL CAHTEKKDKA KEDLRLALKE GHSVLESLRE LQAEGSEPSV NQDQLDNQAT VQRLLAQLNE TEAAFDEFWA KHQQKLEQCL
This gene encodes a guanine nucleotide exchange factor that interacts specifically with the GTP-bound Rac1 and plays a role in the Rho/Rac signaling pathways. A variant in this gene was associated with osteoarthritis. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DBL's big sister; FLJ12122; Guanine nucleotide exchange factor DBS; MCF2 transforming sequence-like protein; MCF2-transforming sequence-like protein; RP11-98F14.2
Gene Aliases: ARHGEF14; DBS; KIAA0362; MCF2L; OST
UniProt ID: (Human) O15068
Entrez Gene ID: (Human) 23263
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support