Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSSPASTPSR RGSRRGRATP AQTPRSEDAR SSPSQRRRGE DSTSTGELQP MPTSPGVDLQ SPAAQDVLFS SPPQMHSSAI PLDFDVSSPL TYGTPSSRVE GTPRSGVRGT PVRQRPDLGS AQKGLQVDLQ SDGAAAEDIV ASEQSLGQKL VIWGTDVNVA ACKENFQRFL QRFIDPLAKE EENVGIDITE PLYMQRLGEI NVIGEPFLNV NCEHIKSFDK NLYRQLISYP QEVIPTFDMA VNEIFFDRYP DSILEHQIQV RPFNALKTKN MRNLNPEDID QLITISGMVI RTSQLIPEMQ EAFFQCQVCA HTTRVEMDRG RIAEPSVCGR CHTTHSMALI HNRSLFSDKQ MIKLQESPED MPAGQTPHTV ILFAHNDLVD KVQPGDRVNV TGIYRAVPIR VNPRVSNVKS VYKTHIDVIH YRKTDAKRLH GLDEEAEQKL FSEKRVELLK ELSRKPDIYE RLASALAPSI YEHEDIKKGI LLQLFGGTRK DFSHTGRGKF RAEINILLCG DPGTSKSQLL QYVYNLVPRG QYTSGKGSSA VGLTAYVMKD PETRQLVLQT GALVLSDNGI CCIDEFDKMN ESTRSVLHEV MEQQTLSIAK AGIICQLNAR TSVLAAANPI ESQWNPKKTT IENIQLPHTL LSRFDLIFLM LDPQDEAYDR RLAHHLVALY YQSEEQAEEE LLDMAVLKDY IAYAHSTIMP RLSEEASQAL IEAYVDMRKI GSSRGMVSAY PRQLESLIRL AEAHAKVRLS NKVEAIDVEE AKRLHREALK QSATDPRTGI VDISILTTGM SATSRKRKEE LAEALKKLIL SKGKTPALKY QQLFEDIRGQ SDIAITKDMF EEALRALADD DFLTVTGKTV RLL
The protein encoded by this gene is one of the highly conserved mini-chromosome maintenance proteins (MCM) that are essential for the initiation of eukaryotic genome replication. The hexameric protein complex formed by MCM proteins is a key component of the pre-replication complex (pre_RC) and may be involved in the formation of replication forks and in the recruitment of other DNA replication related proteins. The MCM complex consisting of this protein and MCM2, 6 and 7 proteins possesses DNA helicase activity, and may act as a DNA unwinding enzyme. The phosphorylation of this protein by CDC2 kinase reduces the DNA helicase activity and chromatin binding of the MCM complex. This gene is mapped to a region on the chromosome 8 head-to-head next to the PRKDC/DNA-PK, a DNA-activated protein kinase involved in the repair of DNA double-strand breaks. Alternatively spliced transcript variants encoding the same protein have been reported.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CDC21 homolog; DNA replication licensing factor MCM4; homolog of S. pombe cell devision cycle 21; mini chromosome maintenance deficient 4 homolog; minichromosome maintenance deficient 4; P1-CDC21
Gene Aliases: 19G; AI325074; AU045576; CDC21; CDC54; hCdc21; mcdc21; MCM4; Mcmd4; mKIAA4003; NKCD; NKGCD; P1-CDC21
UniProt ID: (Human) P33991, (Mouse) P49717
Entrez Gene ID: (Human) 4173, (Mouse) 17217
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support