Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MALKDYALEK EKVKKFLQEF YQDDELGKKQ FKYGNQLVRL AHREQVALYV DLDDVAEDDP ELVDSICENA RRYAKLFADA VQELLPQYKE REVVNKDVLD VYIEHRLMME QRSRDPGMVR SPQNQYPAEL MRRFELYFQG PSSNKPRVIR EVRADSVGKL VTVRGIVTRV SEVKPKMVVA TYTCDQCGAE TYQPIQSPTF MPLIMCPSQE CQTNRSGGRL YLQTRGSRFI KFQEMKMQEH SDQVPVGNIP RSITVLVEGE NTRIAQPGDH VSVTGIFLPI LRTGFRQVVQ GLLSETYLEA HRIVKMNKSE DDESGAGELT REELRQIAEE DFYEKLAASI APEIYGHEDV KKALLLLLVG GVDQSPRGMK IRGNINICLM GDPGVAKSQL LSYIDRLAPR SQYTTGRGSS GVGLTAAVLR DSVSGELTLE GGALVLADQG VCCIDEFDKM AEADRTAIHE VMEQQTISIA KAGILTTLNA RCSILAAANP AYGRYNPRRS LEQNIQLPAA LLSRFDLLWL IQDRPDRDND LRLAQHITYV HQHSRQPPSQ FEPLDMKLMR RYIAMCREKQ PMVPESLADY ITAAYVEMRR EAWASKDATY TSARTLLAIL RLSTALARLR MVDVVEKEDV NEAIRLMEMS KDSLLGDKGQ TARTQRPADV IFATVRELVS GGRSVRFSEA EQRCVSRGFT PAQFQAALDE YEELNVWQVN ASRTRITFV
MCM7 encodes a protein that is one of the highly conserved mini-chromosome maintenance proteins (MCM) that are essential for the initiation of eukaryotic genome replication. The hexameric protein complex formed by the MCM proteins is a key component of the pre-replication complex (pre_RC) and may be involved in the formation of replication forks and in the recruitment of other DNA replication related proteins. The MCM complex consisting of this protein and MCM2, 4 and 6 proteins possesses DNA helicase activity, and may act as a DNA unwinding enzyme.The protein encoded by this gene is one of the highly conserved mini-chromosome maintenance proteins (MCM) that are essential for the initiation of eukaryotic genome replication. The hexameric protein complex formed by the MCM proteins is a key component of the pre-replication complex (pre_RC) and may be involved in the formation of replication forks and in the recruitment of other DNA replication related proteins. The MCM complex consisting of this protein and MCM2, 4 and 6 proteins possesses DNA helicase activity, and may act as a DNA unwinding enzyme. Cyclin D1-dependent kinase, CDK4, is found to associate with this protein, and may regulate the binding of this protein with the tumorsuppressor protein RB1/RB. Alternatively spliced transcript variants encoding distinct isoforms have been reported.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CDC47 homolog; DNA replication licensing factor MCM7; homolog of S. cerevisiae Cdc47; minichromosome maintenance deficient 7; P1 P85MCM; P1.1-MCM3; protein phosphatase 1, regulatory subunit 104
Gene Aliases: CDC47; MCM2; MCM7; P1.1-MCM3; P1CDC47; P85MCM; PNAS146; PPP1R104
UniProt ID: (Human) P33993
Entrez Gene ID: (Human) 4176
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support