Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MNSDQVTLVG QVFESYVSEY HKNDILLILK ERDEDAHYPV VVNAMTLFET NMEIGEYFNM FPSEVLTIFD SALRRSALTI LQSLSQPEAV SMKQNLHARI SGLPVCPELV REHIPKTKDV GHFLSVTGTV IRTSLVKVLE FERDYMCNKC KHVFVIKADF EQYYTFCRPS SCPSLESCDS SKFTCLSGLS SSPTRCRDYQ EIKIQEQVQR LSVGSIPRSM KVILEDDLVD SCKSGDDLTI YGIVMQRWKP FQQDVRCEVE IVLKANYIQV NNEQSSGIIM DEEVQKEFED FWEYYKSDPF AGRNVILASL CPQVFGMYLV KLAVAMVLAG GIQRTDATGT RVRGESHLLL VGDPGTGKSQ FLKYAAKITP RSVLTTGIGS TSAGIVCDNF K
Component of the MCM8-MCM9 complex, a complex involved in homologous recombination repair following DNA interstrand cross-links and plays a key role during gametogenesis. The MCM8- MCM9 complex probably acts as a hexameric helicase downstream of the Fanconi anemia proteins BRCA2 and RAD51 and is required to process aberrant forks into homologous recombination substrates and to orchestrate homologous recombination with resection, fork stabilization and fork restart.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA helicase MCM9; DNA replication licensing factor MCM9; hMCM9; Mini-chromosome maintenance deficient domain-containing protein 1; Minichromosome maintenance 9; minichromosome maintenance complex component 9
Gene Aliases: C6orf61; dJ329L24.1; dJ329L24.3; MCM9; MCMDC1; ODG4; RGD1560557
UniProt ID: (Human) Q9NXL9
Entrez Gene ID: (Human) 254394, (Rat) 499437
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support