Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
MA1-40041 detects MD-1 from mouse samples.
MA1-40041 has been successfully used in flow cytometry and functional assays applications.
The MA1-40041 immunogen is mouse MD-1 extracellular protein.
MD-1 is an extracellular protein associated with the extracellular domain of RP105.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ly-86; Lymphocyte antigen 86; Protein MD-1; RP1-80N2.1
Gene Aliases: Ly86; MD-1; MD1
UniProt ID: (Mouse) O88188
Entrez Gene ID: (Mouse) 17084
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support