Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bethyl Laboratories
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Predicted reactivity by homology: Porcine, Rat, Mouse.
MEA1 gene ontology annotations related to this gene include spermatogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Male-enhanced antigen (H-Y structural gene); Male-enhanced antigen 1; MEA-1
Gene Aliases: HYS; MEA; MEA1
UniProt ID: (Human) Q16626
Entrez Gene ID: (Human) 4201
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support