Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Antibody detects endogenous levels of total MED22.
This gene is located in the surfeit gene cluster, a group of very tightly linked housekeeping genes that do not share sequence similarity. The gene is oriented in a head-to-head fashion with RPL7A (SURF3) and the two genes share a bidirectional promoter. The encoded proteins are localized to the cytoplasm. Two alternative transcript variants encoding different isoforms have been identified for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Mediator complex subunit 22; Mediator of RNA polymerase II transcription subunit 22; MGC48682; Surf-5; surfeit 5; surfeit gene 5; Surfeit locus protein 5
Gene Aliases: AW212655; AW558812; MED22; MED24; RGD1564893; Surf-5; SURF5
UniProt ID: (Human) Q15528, (Rat) A0JPN6, (Mouse) Q62276
Entrez Gene ID: (Human) 6837, (Rat) 499762, (Mouse) 20933
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support