Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MGEPQQVSAL PPPPMQYIKE YTDENIQEGL APKPPPPIKD SYMMFGNQFQ CDDLIIRPLE SQGIERLHPM QFDHKKELRK LNMSILINFL DLLDILIRSP GSIKREEKLE DLKLLFVHVH HLINEYRPHQ ARETLRVMME VQKRQRLETA ERFQKHLERV IEMIQNCLAS LPDDLPHSEA GMRVKTEPMD ADDSNNCTGQ NEHQRENSGH RRDQIIEKDA ALCVLIDEMN ERP
The activation of gene transcription is a multistep process that is triggered by factors that recognize transcriptional enhancer sites in DNA. These factors work with co-activators to direct transcriptional initiation by the RNA polymerase II apparatus. The protein encoded by this gene is a subunit of the CRSP (cofactor required for SP1 activation) complex, which, along with TFIID, is required for efficient activation by SP1. This protein is also a component of other multisubunit complexes e.g. thyroid hormone receptor-(TR-) associated proteins which interact with TR and facilitate TR function on DNA templates in conjunction with initiation factors and cofactors. Two transcript variants encoding the same protein have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Activator-recruited cofactor 34 kDa component; ARC34; Cofactor required for Sp1 transcriptional activation subunit 9; cofactor required for Sp1 transcriptional activation, subunit 9 (33kD); cofactor required for Sp1 transcriptional activation, subunit 9, 33kDa; CRSP complex subunit 9; hMED7; Mediator complex subunit 7; Mediator of RNA polymerase II transcription subunit 7; MGC12284; RNA polymerase transcriptional regulation mediator subunit 7 homolog; Transcriptional coactivator CRSP33
Gene Aliases: ARC34; CRSP33; CRSP9; MED7
UniProt ID: (Human) O43513
Entrez Gene ID: (Human) 9443
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support