Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Active Motif
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
These gene encode homeobox proteins belonging to the TALE family of homeodomain-containing proteins. TALE homeobox proteins are highly conserved transcription regulators, and several members have been shown to be essential contributors to developmental programs. Multiple transcript variants encoding distinct isoforms have been described for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cbp/p300-interacting transactivator with Glu/Asp-rich carboxy-terminal domain 2; Homeobox protein Meis1; Homeobox protein Meis2; Homeobox protein Meis3; leukemogenic homolog protein; Meis homolog 2; Meis1, myeloid ecotropic viral integration site 1 homolog; Meis1, myeloid ecotropic viral integration site 1 homolog 2; Meis1, myeloid ecotropic viral integration site 1 homolog 3; Meis1-related gene 1; Meis1-related protein 1; Meis1-related protein 2; Meis2 homeobox; Myeloid ecotropic viral integration site 1; myeloid ecotropic viral integration site-related gene 1; TALE homeobox protein Meis2; WUGSC:H_NH0444B04.1
Gene Aliases: A430109D20Rik; AI573393; C530044H18Rik; Evi8; HsT18361; MEIS1; MEIS2; MEIS3; MRG1; MRG2; Stra10
UniProt ID: (Human) O00470, (Human) O14770, (Human) Q99687, (Mouse) Q60954, (Mouse) P97367, (Mouse) P97368
Entrez Gene ID: (Human) 4211, (Human) 4212, (Human) 56917, (Mouse) 17268, (Mouse) 17536, (Mouse) 17537
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support