Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • Menin Antibodies

          Abnova

          MEN1 Polyclonal Antibody, MaxPab™

          View all (21) Menin antibodies
          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite MEN1 Polyclonal Antibody, MaxPab™

          • Antibody Testing Data (1)
          MEN1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          MEN1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          MEN1 Antibody (H00004221-B01P) in WB

          MEN1 MaxPab polyclonal antibody. Western Blot analysis of MEN1 expression in Jurkat. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          MEN1 Antibody in Western Blot (WB)

          Product Details

          H00004221-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          Immunocytochemistry (ICC/IF)

          10-100 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          MEN1 (AAH02544.1, 1 a.a. approximately 575 a.a) full-length human protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.57 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Wet ice

          Product Specific Information

          Product may be used with Western Blot (Cell lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MGLKAAQKTL FPLRSIDDVV RLFAAELGRE EPDLVLLSLV LGFVEHFLAV NRVIPTNVPE LTFQPSPAPD PPGGLTYFPV ADLSIIAALY ARFTAQIRGA VDLSLYPREG GVSSRELVKK VSDVIWNSLS RSYFKDRAHI QSLFSFITGT KLDSSGVAFA VVGACQALGL RDVHLALSED HAWSWLYLKG SYMRCDRKME VAFMVCAINP SIDLHTDSLE LLQLQQKLLW LLYDLGHLER YPMALGNLAD LEELEPTPGR PDPLTLYHKG IASAKTYYRD EHIYPYMYLA GYHCRNRNVR EALQAWADTA TVIQDYNYCR EDEEIYKEFF EVANDVIPNL LKEAASLLEA GEERPGEQSQ GTQSQGSALQ DPECFAHLLR FYDGICKWEE GSPTPVLHVG WATFLVQSLG RFEGQVRQKV RIVSREAEAA EAEEPWGEEA REGRRRGPRR ESKPEEPPPP KKPALDKGLG TGQGAVSGPP RKPPGTVAGT ARGPEGGSTA QVPAPAASPP PEGPVLTFQS EKMKGMKELL VATKINSSAI KLQLTAQSQV QMKKQKVSTP SDYTLSFLKR QRKGL

          Target Information

          Potassium channels are a group of ubiquitously expressed proteins that serve numerous functions in excitable and non-excitable cells. One class of integral membrane potassium channels is the large conductance, calcium-activated potassium channel (Maxi K+). Maxi K+ differs from most other potassium channels in that its activation is controlled by both increases in intracellular calcium and by membrane depolarization. Maxi K+ dual activation is possible because of its structure. The core of the channel, which is similar to other potassium channels, is a Maxi K+ alpha homotetramer that contains both a voltage sensor and an intracellular calcium binding domain. In vascular smooth muscle, an auxiliary beta-subunit is found in a 1:1 stoichiometry. The beta-subunit exhibits its effect on the Maxi K+ channel by effectively decreasing by 5- to 10- fold the concentration of calcium required to keep the pore open.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Menin; multiple endocrine neoplasia I

          View more View less

          Gene Aliases: MEAI; MEN1; SCG2

          View more View less

          UniProt ID: (Human) O00255

          View more View less

          Entrez Gene ID: (Human) 4221

          View more View less

          Function(s)
          four-way junction DNA binding Y-form DNA binding chromatin binding double-stranded DNA binding protein binding histone-lysine N-methyltransferase activity protein binding, bridging transcription regulatory region DNA binding protein N-terminus binding R-SMAD binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter MAPK cascade mitotic cell cycle negative regulation of protein phosphorylation osteoblast development type B pancreatic cell differentiation DNA repair transcription, DNA-templated cellular response to DNA damage stimulus brain development negative regulation of cell proliferation response to UV response to gamma radiation negative regulation of cell-substrate adhesion positive regulation of transforming growth factor beta receptor signaling pathway positive regulation of protein binding regulation of activin receptor signaling pathway histone lysine methylation negative regulation of sequence-specific DNA binding transcription factor activity negative regulation of osteoblast differentiation negative regulation of cyclin-dependent protein serine/threonine kinase activity negative regulation of cell cycle negative regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter negative regulation of JNK cascade decidualization negative regulation of epithelial cell proliferation negative regulation of telomerase activity regulation of type B pancreatic cell proliferation cellular response to glucose stimulus cellular response to peptide hormone stimulus response to transforming growth factor beta negative regulation of cell cycle G1/S phase transition beta-catenin-TCF complex assembly
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7fd48b9cb8-69xq6:80/100.66.134.130:80.
          git-commit: fe0b5dd8e7a67b57f27a6e2d26c18f8954d91d59
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0