Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MEHPLFGCL RSPHATAQGL HPFSQSSLAL HGRSDHMSYP ELSTSSSSCI IAGYPNEEGM FASQHHRGHH HHHHHHHHHH QQQQHQALQT NWHLPQMSSP PSAARHSLCL QPDSGGPPEL GSSPPVLCSN SSSLGSSTPT GAACAPGDYG RQALSPAEAE KRSGGKRKSD SSDSQEGNYK SEVNSKPRKE RTAFTKEQIR ELEAEFAHHN YLTRLRRYEI AVNLDLTERQ VKVWFQNRRM KWKRVKGGQQ GAAAREKELV NVKKGTLLPS ELSGIGAATL QQTGDSIANE DSHDSDHSSE HAHL (1-303 aa encoded by BC017021)
Mesodermal transcription factor that plays a key role in somitogenesis and is required for sclerotome development. Activates expression of CDKN1A and CDKN2A in endothelial cells, acting as a regulator of vascular cell proliferation. While it activates CDKN1A in a DNA-dependent manner, it activates CDKN2A in a DNA-independent manner. May have a regulatory role when quiescent vascular smooth muscle cells reenter the cell cycle.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Growth arrest-specific homeobox; Homeobox protein MOX-2; mesenchyme homeo box 2 (growth arrest-specific homeo box); Mesenchyme homeobox 2
Gene Aliases: AI528662; GAX; MEOX2; Mox-2; MOX2
UniProt ID: (Human) P50222, (Rat) P39020, (Mouse) P32443
Entrez Gene ID: (Human) 4223, (Rat) 29279, (Mouse) 17286
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support