Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAKLHIPVMV DEVVHCLSPQ KGQIFLDMTF GSGGHTKAIL QKESDIVLYA LDRDPTAYAL AEHLSELYPK QIRAMLGQFS QAEALLMKAG VQPGTFDGVL MDLGCSSMQL DTPERGFSLR KDGSLDMRMD GGRSTGTCIY PKNIRGGEAC QENRFSNCSG TQHLPHHQNP AACQHRCRSI SSLCYLYTER LTTAIYPYCH QDFPGSSHIC EQ
METT5D1 belongs to the methyltransferase superfamily, mraW family. METT5D1 is a probable S-adenosyl-L-methionine-dependent methyltransferase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 12S rRNA m4C methyltransferase; 12S rRNA N(4)-cytidine methyltransferase METTL15; methyltransferase 5 domain containing 1; Methyltransferase 5 domain-containing protein 1; Methyltransferase-like protein 15; probable S-adenosyl-L-methionine-dependent methyltransferase METT5D1; putative S-adenosyl-L-methionine-dependent methyltransferase METT5D1
Gene Aliases: METT5D1; METTL15
UniProt ID: (Human) A6NJ78
Entrez Gene ID: (Human) 196074
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support