Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSSGADGGGG AAVAARSDKG SPGEDGFVPS ALGTREHWDA VYERELQTFR EYGDTGEIWF GEESMNRLIR WMQKHKIPLD ASVLDIGTGN GVFLVELAKF GFSNITGIDY SPSAIQLSGS IIEKEGLSNI KLKVEDFLNL STQLSGFHIC IDKGTFDAIS LNPDNAIEKR KQYVKSLSRV LKVKGFFSNN VM
Protein-lysine methyltransferase that selectively catalyzes the trimethylation of EEF1A at 'Lys-318'. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EEF1A lysine methyltransferase 2; Em:AC068896.3; Methyltransferase-like protein 10; Protein-lysine N-methyltransferase METTL10
Gene Aliases: C10orf138; EEF1AKMT2; METTL10
UniProt ID: (Human) Q5JPI9
Entrez Gene ID: (Human) 399818
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support