Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MFAP3L is a protein coding gene. May participate in the nuclear signaling of EGFR and MAPK1/ERK2. May a have a role in metastasis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: microfi brillar-associated protein 3-like; Microfibrillar-associated protein 3-like; Testis development protein NYD-SP9
Gene Aliases: 4933428A15Rik; 5430405D20Rik; AI461995; AW125052; HSD-39; HSD39; KIAA0626; MFAP3L; mKIAA0626; NYD-sp9
UniProt ID: (Human) O75121, (Mouse) Q9D3X9, (Rat) Q6AYP2
Entrez Gene ID: (Human) 9848, (Mouse) 71306, (Rat) 306424
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support