Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%
MFSD1 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include integral component of membrane.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Lysosomal dipeptide transporter MFSD1; Major facilitator superfamily domain-containing protein 1; SMAP-4; Smooth muscle cell-associated protein 4
Gene Aliases: MFSD1; SMAP4; UG0581B09
UniProt ID: (Human) Q9H3U5
Entrez Gene ID: (Human) 64747
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support