Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: PGSTEKGLPLR KTHVLLGRSH DTVQEAATSR RARASKKTAQ PWVEVVLALR NMKNLLF
Highest antigen sequence identity to the following orthologs - mouse 41%, rat 38%.
MFSD9 gene ontology annotations related to this gene include transmembrane transport.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Major facilitator superfamily domain-containing protein 9
Gene Aliases: MFSD9; PSEC0066
UniProt ID: (Human) Q8NBP5
Entrez Gene ID: (Human) 84804
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support