Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • MGP Antibodies

          Proteintech

          MGP Monoclonal Antibody (1A1C3), CoraLite® Plus 488

          View all (21) MGP antibodies

          Promotion

          View available promotion(s)

          Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite MGP Monoclonal Antibody (1A1C3), CoraLite® Plus 488

          • Antibody Testing Data (1)
          MGP Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          MGP Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          MGP Antibody (CL488-60055) in ICC/IF

          Immunofluorescent analysis of (4% PFA) fixed MDA-MB-231 cells using CoraLite® Plus 488 MGP antibody (CL488-60055, Clone: 1A1C3 ) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          MGP Antibody in Immunocytochemistry (ICC/IF)

          Product Details

          CL488-60055

          Applications
          Tested Dilution
          Publications

          Immunocytochemistry (ICC/IF)

          1:50-1:500
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Mouse / IgG2a

          Class

          Monoclonal

          Type

          Antibody

          Clone

          1A1C3

          Immunogen

          Recombinant protein
          View immunogen

          Conjugate

          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation

          Excitation/Emission Max

          493/522 nm

          Form

          Liquid

          Purification

          Protein A

          Storage buffer

          PBS with 50% glycerol, 0.5% BSA

          Contains

          0.05% ProClin 300

          Storage conditions

          -20°C, store in dark

          Shipping conditions

          Wet ice

          Product Specific Information

          Immunogen sequence: MKSLILLAIL AALAVVTLCY ESHESMESYE LNPFINRRNA NTFISPQQRW RAKVQERIRE RSKPVHELNR EACDDYRLCE RYAMVYGYNA AYNRYFRKRR GAK

          Target Information

          MGP is associates with the organic matrix of bone and cartilage. MGP is thought to act as an inhibitor of bone formation.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Cell growth-inhibiting gene 36 protein; MATRIX; matrix gamma-carboxyglutamate (gla) protein; Matrix Gla protein; MGP

          View more View less

          Gene Aliases: GIG36; MGLAP; MGP; NTI

          View more View less

          UniProt ID: (Human) P08493, (Mouse) P19788

          View more View less

          Entrez Gene ID: (Human) 4256, (Mouse) 17313

          View more View less

          Function(s)
          extracellular matrix structural constituent calcium ion binding protein binding structural constituent of bone calcium-dependent protein binding
          Process(es)
          cartilage condensation ossification cell differentiation regulation of bone mineralization protein complex assembly multicellular organism development lung development branching morphogenesis of an epithelial tube cartilage development
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5d94444789-f7m9l:80/100.66.129.252:80.
          git-commit: ec3573d8beaf0fd412f2d0ea2dcb8dd14c74098c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0