Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Like their mammalian counterparts, avian MHC Class I molecules, also known as B-F antigens, consist of a highly polymorphic alpha-chain noncovalently bound to the invariant beta2-microglobulin subunit. MHC Class I molecules are expressed on most nucleated cells where they present endogenously synthesized antigenic peptides to CD8+ T lymphocytes, which are usually cytotoxic T cells. The monoclonal antibody F21-2 also reacts with turkey MHC Class I.
HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B-F histocompatibility F10 antigen; B-F major; B-F minor; B-F-beta-IV; B12; class I histocompatibility antigen, F10 alpha chain; F10 alpha chain; Major histocompatibility complex class I antigen BF2; major histocompatibility complex class I glycoprotein; major histocompatibility complex, class I, G; MHC BF1 class I; MHC BF2 class I; MHC class I alpha chain 1; MHC class I alpha chain 2; MHC class I antigen alpha chain; MHC class I antigen B-F heavy chain; MHC class I antigen B-F major heavy chain; MHC class I glycoprotein; MHC class I molecule
Gene Aliases: B-F; B-F-S01; B-F-S02; B-F-S03; B-F-S04; B-F-S05; B-F-S06; B-F-S07; B-F-S09; B-F-S11; B-F-S12; B-F-S13; B-F-S14; B-FI; B-FIV; B-LB2; BF; BF12; BF2; BFa1; BFa2; BFIV; BFIV21; BFw-03; BFw-05; BFz-01
Entrez Gene ID: (Chicken) 693260, (Chicken) 425389
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support