Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MMQICDTYN QKHSLFNAMN RFIGAVNNMD QTVMVPSLLR DVPLADPGLD NDVGVEVGGS GGCLEERTPP VPDSGSANGS FFAPSRDMYS HYVLLKSIRN DIEWGVLHQP PPPAGSEEGS AWKSKDILVD LGHLEGADAG EEDLEQQFHY HLRGLHTVLS KLTRKANILT NRYKQEIGFG NWGH (1-183 aa encoded by BC008908)
Plays a role in the regulation of lipogenesis in liver. Up-regulates ACACA enzyme activity. Required for efficient lipid biosynthesis, including triacylglycerol, diacylglycerol and phospholipid. Involved in stabilization of microtubules.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ10386; gastrulation specific G12 homolog; Gastrulation-specific G12-like protein; MID1 interacting G12-like protein; MID1 interacting protein 1 (gastrulation specific G12 homolog); MID1 interacting protein 1 (gastrulation specific G12-like); Mid1-interacting G12-like protein; Mid1-interacting protein 1; mid1ip1 {ECO:0000250|UniProtKB:Q9CQ20}; Protein STRAIT11499; Protein STRAIT11499 homolog; S14R; spot 14-like androgen-inducible protein; spot 14-R; Spot 14-related protein
Gene Aliases: 3110038L01Rik; G12-like; MID1IP1; MIG12; S14R; slap; STRAIT11499; THRSPL
UniProt ID: (Human) Q9NPA3, (Mouse) Q9CQ20, (Rat) Q6P7D5
Entrez Gene ID: (Human) 58526, (Mouse) 68041, (Rat) 404280
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support