Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MEPQPGGARS CRRGAPGGAC ELGPAAEAAP MSLAIHSTTG TRYDLAVPPD ETVEGLRKRL SQRLKVPKER LALLHKDTRL SSGKLQEFGV GDGSKLTLVP TVEAGLMSQA SRPEQSVMQA LESLTETQVS DFLSGRSPLT LALRVGDHMM FVQLQLAAQH APLQHRHVLA AAAAAAAARG DPSIASPVSS PCRPVSSAAR VPPVPTSPSP ASPSPITAGS FRSHAASTTC PEQMDCSPTA SSSASPGAST TSTPGASPAP RSRKPGAVIE SFVNHAPGVF SGTFSGTLHP NCQDSSGRPR RDIGTILQIL NDLLSATRHY QGMPPSLAQL RCHAQCSPAS PAPDLAPRTT SCEKLTAAPS ASLLQGQSQI RMCKPPGDRL RQTENRATRC KVERLQLLLQ QKRLRRKARR DARGPYHWSP SRKAGRSDSS SSGGGGSPSE ASGLGLDFED SVWKPEANPD IKSEFVVA
Expression of the midnolin gene is developmentally regulated: it is strongly expressed at the mesencephalon (midbrain) of the embryo in day 12.5 (E12.5) mice. The midnolin encodes a protein of 508 amino acids (aa), which contains a Ubiquitin-like domain. The intracellular distribution of the midnolin was studied by using midnolin-green fluorescent protein (GFP) fusion proteins. Midnolin was found to be localized in the nucleus and nucleolus, but not in the cytoplasm. The nucleolar localization signal was determined to be a 28aa peptide located at the C-terminal region of the midnolin. May be involved in regulation of genes related to neurogenesis in the nucleolus.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp547M072; Midbrain nucleolar protein; Midnolin
Gene Aliases: MIDN
UniProt ID: (Human) Q504T8
Entrez Gene ID: (Human) 90007
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support