Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: DQLLWCPDV VLESKVKEYL VETSLRTGSE KIMDRISAGT HTRDNEQALY ELLKCNHNIK EAIERYCCNG KASQGMTAWT EEECRSFEHA LMLFGKDFHL IQKNKVRTRT VAECVAFYYM WKKSERYDYF AQQTRFGKKR YNHHPGVTDY MDRLVDETEA LGGTVNASAL TSNRPEPIPD QQLNILNSFT ASDLTALTNS VATVCDPTDV NCLDDSFPPL GNTPRGQVNH VPVVTEELLT LPSNGESDCF NLFETGFYHS ELNPMNMCSE ESERPAKRLK MGIAVPESFM NEVSVNNLGV DFENHTHHIT SAKMAVSVAD FGSLSANETN GFISAHALHQ HAALHSE (177-522 aa encoded by BC041348 )
MIER3, known as Mesoderm Induction Early Response 1, Family Member 3, is a gene associated with epithelial-mesenchymal transition (EMT) processes particularly in cancer research. The gene is located on chromosome 7 and has been identified in studies as playing a crucial role in the progression of colorectal and breast cancers. Structurally, MIER3 is involved in chromatin remodeling and histone acetylation, functioning together with HDAC1/2 and Snail proteins. Its expression profile has been characterized using bioinformatics, real-time PCR, and Western blotting, indicating that MIER3 interacts closely with cellular mechanisms that modulate gene expression and cellular differentiation. The gene's role in cancer has been explored through its capacity to induce EMT, which affects tumor metastasis and cell migration, highlighting its potential as a therapeutic target in oncology.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: mesoderm induction early response 1, family member 3; Mesoderm induction early response protein 3; Mi-er3
Gene Aliases: 5730509D14; AU015422; D130064H19Rik; MIER3; RGD1311241
UniProt ID: (Human) Q7Z3K6, (Mouse) Q3UHF3
Entrez Gene ID: (Human) 166968, (Rat) 310086, (Mouse) 218613
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support