Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: SISYSCSAVPH QGRGFSQYGV SGSPTKSKVT SCPGCRKPLP RCALCLINMG TPVSSCPGGT KSDEKVDLSK DKKLAQFNNW FTWCHNCRHG GHAGHMLSWF RDHAECPVSA CTCKCMQLD
Highest antigen sequence identity to the following orthologs - mouse 98%, rat 98%.
As a component of the GATOR subcomplex GATOR2, functions within the amino acid-sensing branch of the TORC1 signaling pathway. Indirectly activates mTORC1 and the TORC1 signaling pathway through the inhibition of the GATOR1 subcomplex (PubMed:23723238). It is negatively regulated by the upstream amino acid sensors SESN2 and CASTOR1 (PubMed:25457612, PubMed:27487210). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ20323; GATOR2 complex protein MIOS; H_DJ1159O04.1; Missing oocyte meiosis regulator homolog; missing oocyte, meiosis regulator, homolog; WUGSC:H_DJ1159O04.1
Gene Aliases: MIO; MIOS; Sea4
UniProt ID: (Human) Q9NXC5
Entrez Gene ID: (Human) 54468
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support