Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MSVDPMTYEA QFFGFTPQTC MLRIYIAFQG YLFEVMQAVE QVILRKLDGI PDCDISPVQI RKCTEKFLCF MKGHFDNLFS KMEQLFLQLI LRIPSNILLP EDKCKETPYS EEDFQHLQKE IEQLQEKYKT ELCTKQALLA ELEEQKIVQA KLKQTLTFFD ELHNVGRDHG TSDFRESLVS LVQNSRKLQN IRDNVEKESK RLKIS
This protein is part of the MIS12 complex which is required for normal chromosome alignment and segregation and for kinetochore formation during mitosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: homolog of yeast Mis12; MIS12 homolog; MIS12 kinetochore complex component; MIS12, MIND kinetochore complex component, homolog; Protein MIS12 homolog
Gene Aliases: 2510025F08Rik; hMis12; KNTC2AP; MIS12; MTW1
UniProt ID: (Human) Q9H081
Entrez Gene ID: (Human) 79003
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support