Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: MGAVNPLLSQ AESSHTEPDL EDCSFRCRGT SPQESLSSMS PISSLPALFD QTASAPCGGG QLDPAAPGTT NMEQLLEKQG DGEAGVNIVE MLKALHALQK ENQRLQEQIL SLTAKKERLQ ILNVQLSVPF PALPAALPAA NGPVPGPYGL PPQAGSSDSL STSKSPPGKS SLGLDNSLST SSEDPHSGCP SRSSSSLSFH STPPPLPLLQ QSPATLPLAL PGAPAPLPPQ PQNGLGRAPG AAGLGAMPMA EGLLGGLAGS GGLPLNGLLG GLNGAAAPNP ASLSQAGGAP TLQLPGCLNS LTEQQRHLLQ QQEQQLQQLQ QLLASPQLTP EHQTVVYQMI QQIQQKRELQ RLQMAGGSQL PMASLLAGSS TPLLSAGTPG LLPTASAPPL LPAGALVAPS LGNNTSLMAA AAAAAAVAAA GGPPVLTAQT NPFLSLSGAE GSGGGPKGGT ADKGASANQE KG
MLLT6 gene ontology annotations related to this gene include negative regulation of histone H3-K79 methylation; negative regulation of urine volume; positive regulation of sodium ion transport; positive regulation of transcription by RNA polymerase II; renal potassium excretion; renal sodium excretion.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ALL1-fused gene from chromosome 17 protein; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog); translocated to, 6; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 6; Myeloid/lymphoid or mixed-lineage leukemia, translocated to, 6; myeloid/lymphoid or mixed-lineage leukemia; translocated to, 6; Protein AF-17; trithorax homolog
Gene Aliases: AF17; MLLT6
UniProt ID: (Human) P55198
Entrez Gene ID: (Human) 4302
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support