Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: PPAPSGSERR LSGDLSSMPG PGTLSVRVSP PQPILSRGRP DSNKTENRRI THISAEQKRR FNIKLGFDTL HGLVSTLSAQ PSLKVSKATT LQKTAEYI
ChREBP is a transcription factor involved in activating genes that encode enzymes of fatty acid biosynthesis in liver and adipose. This protein may have relevance to lipid metabolism, obesity, and type II diabetes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bHLHd14; carbohydrate response element binding protein; Carbohydrate-responsive element-binding protein; ChREBP; Class D basic helix-loop-helix protein 14; MLX interacting protein-like; MLX interactor; MLX-interacting protein-like; Williams Beuren syndrome chromosome region 14; Williams-Beuren syndrome chromosomal region 14 protein; Williams-Beuren syndrome chromosome region 14 protein 1; Williams-Beuren syndrome chromosome region 14 protein 2; Williams-Beuren syndrome chromosome region 14 protein 3; WS basic-helix-loop-helix leucine zipper protein; WS-bHLH
Gene Aliases: BHLHD14; CHREBP; MIO; MLXIPL; MONDOB; WBSCR14; WS-bHLH
UniProt ID: (Human) Q9NP71
Entrez Gene ID: (Human) 51085
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support