Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAFPEPKPRP PELPQKRLKT LDCGQGAVRA VRFNVDGNYC LTCGSDKTLK LWNPLRGTLL RTYSGHGYEV LDAAGSFDNS SLCSGGGDKA VVLWDVASGQ VVRKFRGHAG KVNTVQFNEE ATVILSGSID SSIRCWDCRS RRPEPVQTLD EARDGVSSVK VSDHEILAGS VDGRVRRYDL RMGQLFSDYV GSPITCTCFS RDGQCTLVSS LDSTLRLLDK DTGELLGEYK GHKNQEYKLD CCLSERDTHV VSCSEDGKVF FWDLVEGALA LALPVGSGVV QSLAYHPTEP CLLTAMGGSV QCWREEAYEA EDGAG
MORG1 is a member of the WD-40 protein family. The protein is proposed to function as a molecular scaffold for various multimeric protein complexes. The protein associates with several components of the extracellular signal-regulated kinase (ERK) pathway, and promotes ERK activity in response to serum or other signals. The protein also interacts with egl nine homolog 3 (EGLN3, also known as PHD3) and regulates expression of hypoxia-inducible factor 1, and has been purified as part of the spliceosome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MAPK organizer 1; MGC4238; Mitogen-activated protein kinase organizer 1; WD repeat domain-containing protein 83
Gene Aliases: MORG1; WDR83
UniProt ID: (Human) Q9BRX9
Entrez Gene ID: (Human) 84292
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support