Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen sequence: RIRDQAWDDV VRKEKPKEDA YEYKKRLTLD HEKSKLSLAE IYEQEYIKLN QQKTAEEENP EHVEIQKMMD SLFLKLDALS NFHFIPKPPV PEIKVVSNLP AITMEEVAPV SVSDAALLAP EEIKEKNKAG DIKTAAEKTA TDKKRERRKK KYQKRMKIKE KEKRRKLLEK SSVDQAGKYS KTVASEKLKQ LTKTGKASFI KDEGKDKALK SSQAFFSKLQ DQVKMQINDA KKTEKKKKKR QDISVHKLKL; Positive Samples: MCF-7, HeLa; Cellular Location: Chromosome, Nucleus, nucleolus
MPP10 (M-phase phosphoprotein 10), also known as MPHOSPH10, is a 681 amino acid protein that localizes to the fibrillar region of the nucleolus. Phosphorylated in the M (mitotic) phase of the cell cycle, MPP10 is a component of the 60-80S U3 small nucleolar ribonucleoprotein (U3 snoRNP) complex and is required for the early cleavages during pre-18S ribosomal RNA processing. MPP10 forms a heterotrimeric complex with IMP-3 and IMP-4, which may be required for its association with nucleolar components. The gene encoding MPP10 is localized to human chromosome 2, which houses over 500 genes and is the second smallest human chromosome. Mutations in several of the genes that map to chromosome 22 are involved in the development of Phelan-McDermid syndrome, Neurofibromatosis type 2, autism and schizophrenia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: M phase phosphoprotein 10; M-phase phosphoprotein 10 (U3 small nucleolar ribonucleoprotein); protein phosphatase 1, regulatory subunit 106; U3 small nucleolar ribonucleoprotein; U3 small nucleolar ribonucleoprotein protein MPP10
Gene Aliases: 2810453H10Rik; 5730405D16Rik; AI326008; CT90; MPHOSPH10; MPP10; MPP10P; PPP1R106
UniProt ID: (Human) O00566, (Mouse) Q810V0
Entrez Gene ID: (Human) 10199, (Mouse) 67973
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support