Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 3
Sequence of this protein is as follows: MPVLSEDSGL HETLALLTSQ LRPDSNHKEE MGFLRDVFSE KSLSYLMKIH EKLRYYERQS PTPVLHSAVA LAEDVMEELQ AASVHSDERE LLQLLSTPHL RAVLMVHDTV AQKNFDPVLP PLPDNIDEDF DEESVKIVRL VKNKEPLGAT IRRDEHSGAV VVARIMRGGA ADRSGLVHVG DELREVNGIA VLHKRPDEIS QILAQSQGSI TLKIIPATQE EDRLKESKVF MRALFHYNPR EDRAIPCQEA GLPFQRRQVL EVVSQDDPTW WQAKRVGDTN LRAGLIPSKG FQERRLSYRR AAGTLPSPQS LRKPPYDQPC DKETCDCEGY LKGHYVAGLR RSFRLGCRER LGGSQEGKMS SGAESPELLT YEEVARYQHQ PGERPRLVVL IGSLGARLHE LKQKVVAENP QHFGVAVPHT TRPRKSHEKE GVEYHFVSKQ AFEADLHHNK FLEHGEYKEN LYGTSLEAIQ AVMAKNKVCL VDVEPEALKQ LRTSEFKPYI IFVKPAIQEK RKTPPMSPAC EDTAAPFDEQ QQEMAASAAF IDRHYGHLVD AVLVKEDLQG AYSQLKVVLE KLSKDTHWVP VSWVR
This gene product is a member of a family of membrane-associated proteins termed MAGUKs (membrane-associated guanylate kinase homologs). MAGUKs interact with the cytoskeleton and regulate cell proliferation, signaling pathways, and intracellular junctions. This protein contains a conserved sequence, called the SH3 (src homology 3) motif, found in several other proteins that associate with the cytoskeleton and are suspected to play important roles in signal transduction. Alternatively spliced transcript variants have been identified. One transcript variant is experimentally supported, but it doesn't encode a protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Discs large homolog 3; discs, large homolog 3; MAGUK p55 subfamily member 3; membrane protein palmitoylated 3; membrane protein, palmitoylated 3 (MAGUK p55 subfamily member 3); Mpp3 membrane protein, palmitoylated 3; Protein MPP3
Gene Aliases: 6430514B01; DLG3; Dlgh3; MPP3
UniProt ID: (Human) Q13368
Entrez Gene ID: (Human) 4356, (Mouse) 13384
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support