Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAMIELGFGR QNFHPLKRKS SLLLKLIAVV FAVLLFCEFL IYYLAIFQCN WPEVKTTASD GEQTTREPVL KAMFLADTHL LGEFLGHWLD KLRREWQMER AFQTALWLLQ PEVVFILGDI FDEGKWSTPE AWADDVERFQ KMFRHPSHVQ LKVVAGNHDI GFHYEMNTYK VERFEKVFSS ERLFSWKGIN FVMVNSVALN GDGCGICSET EAELIEVSHR LNCSREQARG SSRCGPGPLL PTSAPVLLQH YPLYRRSDAN CSGEDAAPAE ERDIPFKENY DVLSREASQK LLWWLQPRLV LIGHTHSACE VHHGGRVPEL SVPSFSWRNR NNPSFIMGTD A
Metallophosphoesterase required for transport of GPI-anchor proteins from the endoplasmic reticulum to the Golgi. Acts in lipid remodeling steps of GPI-anchor maturation by mediating the removal of a side-chain ethanolamine-phosphate (EtNP) from the second Man (Man2) of the GPI intermediate, an essential step for efficient transport of GPI-anchor proteins. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: metallo phosphoesterase; Metallophosphoesterase 1; Post-GPI attachment to proteins factor 5
Gene Aliases: MPPE1; PGAP5; PP579
UniProt ID: (Human) Q53F39
Entrez Gene ID: (Human) 65258
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support