Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
Positive Samples: HepG2, 293T, HeLa, A-549, BT-474; Cellular Location: Apical cell membrane, Multi-pass membrane protein
Immunogen sequence: IFSAFSENNE SSNNPSSIAS FLSSITYSWY DSIILKGYKR PLTLEDVWEV DEEMKTKTLV SKFETHMKRE LQKARRALQR RQEKSSQQNS GARLPGLNKN QSQSQDALVL EDVEKKKKKS GTKKDVPKSW LMKALF
This gene encodes the prototype of the membrane-associated guanylate kinase (MAGUK) family proteins. MAGUKs interact with the cytoskeleton and regulate cell proliferation, signaling pathways, and intercellular junctions. The encoded protein is an extensively palmitoylated membrane phosphoprotein containing a PDZ domain, a Src homology 3 (SH3) motif, and a guanylate kinase domain. This gene product interacts with various cytoskeletal proteins and cell junctional proteins in different tissue and cell types, and may be involved in the regulation of cell shape, hair cell development, neural patterning of the retina, and apico-basal polarity and tumor suppression pathways in non-erythroid cells. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP-binding cassette sub-family C member 2; ATP-binding cassette, sub-family C (CFTR/MRP), member 2; calicular multispecific organic anion transporter; Canalicular multidrug resistance protein; Canalicular multispecific organic anion transporter 1; Human MRP2; KIAA1010; multidrug resistance associated protein 2; multidrug resistance protein 2; Multidrug resistance-associated protein 2
Gene Aliases: ABC30; ABCC2; AI173996; CMOAT; CMOAT1; CMRP; DJS; MRP2
UniProt ID: (Human) Q92887, (Rat) Q63120, (Mouse) Q8VI47
Entrez Gene ID: (Human) 1244, (Rat) 25303, (Mouse) 12780
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support