Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen sequence: MAGPLQGGGA RALDLLRGLP RVSLANLKPN PGSKKPERRP RGRRRGRKCG RGHKGERQRG TRPRLGFEGG QTPFYIRIPK YGFNEGHSFR RQYKPLSLNR LQYLIDLGRV DPSQPIDLTQ LVNGRGVTIQ PLKRDYGVQL VEEGADTFTA KVNIEVQLAS ELAIAAIEKN GGVVTTAFYD PRSLDIVCKP VPFFLRGQPI PKRMLPPEEL VPYYTDAKNR GYLADPAKFP EARLELARKY GYILPDITKD ELFKMLCTRK DPRQIFFGLA PGWVVNMADK KILKPTDENL LKYYTS
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein that belongs to the EcoL15 ribosomal protein family. A pseudogene corresponding to this gene is found on chromosome 15q.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 39S ribosomal protein L15, mitochondrial; L15mt; Large ribosomal subunit protein uL15m; MRP-L15; ribosomal protein, mitochondrial, L7
Gene Aliases: HSPC145; L15mt; MRP-L15; MRP-L7; MRPL15; RPML7
UniProt ID: (Human) Q9P015, (Mouse) Q9CPR5
Entrez Gene ID: (Human) 29088, (Mouse) 27395
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support