Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAAAMAASSL TVTLGRLASA CSHSILRPSG PGAASLWSAS RRFNSQSTSY LPGYVPKTSL SSPPWPEVVL PDPVEETRHH AEVVKKVNEM IVTGQYGRLF AVVHFASRQW KVTSEDLILI GNELDLACGE RIRLEKVLLV GADNFTLLGK PLLGKDLVRV EATVIEKTES WPRIIMRFRK RKNFKKKRIV TTPQTVLRIN SIEIAPCLL
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. Multiple transcript variants encoding different isoforms were identified through sequence analysis although some may be subject to nonsense-mediated decay (NMD).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 39S ribosomal protein L21, mitochondrial; L21mt; Large ribosomal subunit protein bL21m
Gene Aliases: L21mt; MRP-L21; MRPL21
UniProt ID: (Human) Q7Z2W9
Entrez Gene ID: (Human) 219927
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support