Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MSGTLGKVL CLRNNTIFKQ AFSLLRFRTS GEKPIYSVGG ILLSISRPYK TKPTHGIGKY KHLIKAEEPK KKKGKVEVRA INLGTDYEYG VLNIHLTAYD MTLAESYAQY VHNLCNSLSI KVEESYAMPT KTIEVLQLQD QGSKMLLDSV LTTHERVVQI SGLSATFAEI FLEIIQSSLP EGVRLSVKEH TEEDFKGRFK ARPELEELLA KLK (1-212 aa encoded by BC009764)
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. A pseudogene corresponding to this gene is found on chromosome 6p.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 39S ribosomal protein L48, mitochondrial; L48mt; Large ribosomal subunit protein mL48; MGC13323; MRP-L48
Gene Aliases: 1810030E20Rik; 2610028L11Rik; CGI-118; D4Ertd786e; HSPC290; L48MT; MRP-L48; MRPL48
UniProt ID: (Human) Q96GC5, (Mouse) Q8JZS9
Entrez Gene ID: (Human) 51642, (Mouse) 52443, (Rat) 293149
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support