Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: MAPLGTTVLL WSLLRSSPGV ERVCFRARIQ PWHGGLLQPL PCSFEMGLPR RRFSSEAAES GSPETKKPTF MDEEVQSILT KMTGLNLQKT FKPAIQELKP PTYKLMTQAQ LEEATRQAVE AAKVRLKMPP VLEERVPIND VLAEDKILEG TETTKYVFTD ISYSIPHRER FIVVREPSGT LRKASWEERD RMIQVYFPKE GRKILTPIIF KEENLRTMYS QDRHVDVLNL CFAQFEPDST EYIKVHHKTY EDIDKRGKYD LLRSTRYFGG MVWYFVNNKK IDGLLIDQIQ RDLIDDATNL VQLYHVLHPD GQSAQGAKDQ AAEGINLIKV FAKTEAQKGA YIELTLQTYQ EALSRHSAAS
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein that does not seem to have a counterpart in prokaryotic and fungal-mitochondrial ribosomes. This gene lies telomeric of and is transcribed in the opposite direction from the forkhead box L2 gene. A pseudogene corresponding to this gene is found on chromosome Xq.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 28S ribosomal protein S22, mitochondrial; MRP-S22; S22mt; Small ribosomal subunit protein mS22
Gene Aliases: C3orf5; COXPD5; GIBT; GK002; MRP-S22; MRPS22; RPMS22
UniProt ID: (Human) P82650
Entrez Gene ID: (Human) 56945
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support